missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-4-methyl-3-nitropyridine 98.0+%, TCI America™

Catalog No. A29881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29881G 1 g
A29885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29881G Supplier TCI America Supplier No. A29881G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100367-40-6 | C6H6BrN3O2 | 232.037 g/mol

Chemical Identifiers

CAS 100367-40-6
Molecular Formula C6H6BrN3O2
Molecular Weight (g/mol) 232.037
MDL Number MFCD00092197
InChI Key KFVGEPWMVKZPND-UHFFFAOYSA-N
PubChem CID 7213207
IUPAC Name 5-bromo-4-methyl-3-nitropyridin-2-amine
SMILES CC1=C(C(=NC=C1Br)N)[N+](=O)[O-]

Specifications

Melting Point 170°C
Quantity 1 g
Formula Weight 232.04
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromo-4-methyl-3-nitropyridine