accessibility menu, dialog, popup

UserName

CH3BrNHH2NNOO

CAS RN 100367-40-6

IUPAC Name: 2-amino-5-bromo-4-methyl-3-nitropyridin-1-ium
Synonyms: 2-amino-5-bromo-4-methyl-3-nitropyridin-1-ium
Molecular Weight (g/mol): 233.04
Molecular Formula: C6H7BrN3O2
InChi Key: KFVGEPWMVKZPND-UHFFFAOYSA-O
SMILES: CC1=C(Br)C=[NH+]C(N)=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results