missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-iodopyridine 98.0+%, TCI America™

Catalog No. p-7117972
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25631G 1 g
A25635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25631G Supplier TCI America Supplier No. A25631G
Only null left

Chemical Identifiers

CAS 381233-96-1
Molecular Formula C5H4BrIN2
Molecular Weight (g/mol) 298.91
MDL Number MFCD06659000
InChI Key XPERZSKJGNUSHI-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-3-iodopyridine, 5-bromo-3-iodo-pyridin-2-ylamine, 2-amino-3-iodo-5-bromopyridine, 5-bromo-3-iodo-2-pyridinamine, 2-pyridinamine, 5-bromo-3-iodo, 5-bromo-3-iodo-pyridin-2-amine, pubchem8081, acmc-1csc4, ksc495q4p, 5-bromo-3-iodopyridine-2-amine
PubChem CID 1516508
IUPAC Name 5-bromo-3-iodopyridin-2-amine
SMILES NC1=NC=C(Br)C=C1I

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 1 g
Formula Weight 298.91
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromo-3-iodopyridine