accessibility menu, dialog, popup

UserName

NH2NBrI

CAS RN 381233-96-1

IUPAC Name: 5-bromo-3-iodopyridin-2-amine
Synonyms: 5-bromo-3-iodopyridin-2-amine
Molecular Weight (g/mol): 298.91
Molecular Formula: C5H4BrIN2
InChi Key: XPERZSKJGNUSHI-UHFFFAOYSA-N
SMILES: NC1=NC=C(Br)C=C1I
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)