missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-hydroxypyridine 98.0+%, TCI America™

Catalog No. A26731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26731G 1 g
A26735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26731G Supplier TCI America Supplier No. A26731G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39903-01-0 | C5H5BrN2O | 189.01 g/mol

Chemical Identifiers

CAS 39903-01-0
Molecular Formula C5H5BrN2O
Molecular Weight (g/mol) 189.01
MDL Number MFCD09744143
InChI Key YQADLKDQAXAIKW-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-3-pyridinol, 2-amino-5-bromo-3-hydroxypyridine, 2-amino-3-hydroxy-5-bromopyridine, 3-pyridinol, 2-amino-5-bromo, 2-amino-5-bromo-pyridin-3-ol, 2-amino-5-bromopyridine-3-ol, acmc-209j8w, 2-amino-5-bromopryidin-3-ol, 2-amino-3-hydroxy-5-bromo pyridine, 5-bromo-3-hydroxy-2-amino-pyridine
PubChem CID 11694041
IUPAC Name 2-amino-5-bromopyridin-3-ol
SMILES NC1=NC=C(Br)C=C1O

Specifications

Melting Point 200°C
Quantity 1 g
Formula Weight 189.01
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-bromo-3-hydroxypyridine