accessibility menu, dialog, popup

UserName

NBrNH2OH

CAS RN 39903-01-0

IUPAC Name: 2-amino-5-bromopyridin-3-ol
Synonyms: 2-amino-5-bromopyridin-3-ol
Molecular Weight (g/mol): 189.01
Molecular Formula: C5H5BrN2O
InChi Key: YQADLKDQAXAIKW-UHFFFAOYSA-N
SMILES: NC1=NC=C(Br)C=C1O
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)