missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-3-O-benzyl-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranosyl Azide 98.0+%, TCI America™

Catalog No. A18131G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A18131G Supplier TCI America Supplier No. A18131G

Chemical Identifiers

CAS 80887-27-0
Molecular Formula C22H24N4O5
Molecular Weight (g/mol) 424.457
MDL Number MFCD06797052
InChI Key FVNJPKGJMMREOM-CGXUPHRESA-N
PubChem CID 44629748
IUPAC Name N-[(2R,4aR,6R,7R,8R,8aS)-6-azido-2-phenyl-8-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-7-yl]acetamide
SMILES CC(=O)NC1C(C2C(COC(O2)C3=CC=CC=C3)OC1N=[N+]=[N-])OCC4=CC=CC=C4

Specifications

Color White
Quantity 1 g
Formula Weight 424.46
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-3-O-benzyl-4,6-O-benzylidene-2-deoxy-beta-D-glucopyranosyl Azide
TSCA No
Recommended Storage Freezer