accessibility menu, dialog, popup

UserName

CAS RN 80887-27-0

IUPAC Name: N-[6-azido-8-(benzyloxy)-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-yl]acetamide
Synonyms: N-[6-azido-8-(benzyloxy)-2-phenyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-7-yl]acetamide
Molecular Weight (g/mol): 424.46
Molecular Formula: C22H24N4O5
InChi Key: FVNJPKGJMMREOM-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(OC2COC(OC2C1OCC1=CC=CC=C1)C1=CC=CC=C1)N=[N+]=[N-]
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results