missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3-hydroxypyridine, 95%, Thermo Scientific Chemicals

Catalog No. p-7053937
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6172203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6172203 Supplier Thermo Scientific Chemicals Supplier No. H6172203

2-Amino-5-bromo-3-hydroxypyridine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-bromo-3-hydroxypyridine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 39903-01-0
Molecular Formula C5H5BrN2O
Molecular Weight (g/mol) 189.01
MDL Number MFCD09744143
InChI Key YQADLKDQAXAIKW-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-3-pyridinol, 2-amino-5-bromo-3-hydroxypyridine, 2-amino-3-hydroxy-5-bromopyridine, 3-pyridinol, 2-amino-5-bromo, 2-amino-5-bromo-pyridin-3-ol, 2-amino-5-bromopyridine-3-ol, acmc-209j8w, 2-amino-5-bromopryidin-3-ol, 2-amino-3-hydroxy-5-bromo pyridine, 5-bromo-3-hydroxy-2-amino-pyridine
PubChem CID 11694041
IUPAC Name 2-amino-5-bromopyridin-3-ol
SMILES NC1=NC=C(Br)C=C1O

Specifications

Color White
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 189.01
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Amino-5-bromo-3-hydroxypyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only