missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-1,3,4-thiadiazole, 97%, Thermo Scientific Chemicals

Catalog No. AC448670100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448670100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC448670100 Supplier Thermo Scientific Chemicals Supplier No. 448670100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37566-39-5
Molecular Formula C2H2BrN3S
Molecular Weight (g/mol) 180.02
MDL Number MFCD00464325
InChI Key GLYQQFBHCFPEEU-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-1,3,4-thiadiazole, 5-bromo-1,3,4-thiadiazol-2-ylamine, 2-amino-5-bromo-1,3,4 thiadiazole, 1,3,4-thiadiazol-2-amine, 5-bromo, 5-bromo-1,3,4-thiadiazole-2-amine, 5-bromo-1,3,4-thiadiazole-2-ylamine, 5-bromo-1,3,4 thiadiazol-2-ylamine, bromothiadiazolamine, pubchem20972, ksc496o1t
PubChem CID 3650883
IUPAC Name 5-bromo-1,3,4-thiadiazol-2-amine
SMILES NC1=NN=C(Br)S1

Specifications

Melting Point 167.0°C to 169.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 10 g
Formula Weight 180.03
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromo-1,3,4-thiadiazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only