CAS RN 37566-39-5
IUPAC Name:
5-bromo-1,3,4-thiadiazol-2-amine
Synonyms:
5-bromo-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol):
180.02
Molecular Formula:
C2H2BrN3S
InChi Key:
GLYQQFBHCFPEEU-UHFFFAOYSA-N
SMILES:
NC1=NN=C(Br)S1
1
–
1
of
1
results
1
Promotions Available
2-Amino-5-bromo-1,3,4-thiadiazole, 97%, Thermo Scientific Chemicals
2-Amino-5-bromo-1,3,4-thiadiazole, 97%, Thermo Scientific Chemicals
CAS: 37566-39-5 Molecular Formula: C2H2BrN3S Molecular Weight (g/mol): 180.02 MDL Number: MFCD00464325 InChI Key: GLYQQFBHCFPEEU-UHFFFAOYSA-N Synonym: 2-amino-5-bromo-1,3,4-thiadiazole,5-bromo-1,3,4-thiadiazol-2-ylamine,2-amino-5-bromo-1,3,4 thiadiazole,1,3,4-thiadiazol-2-amine, 5-bromo,5-bromo-1,3,4-thiadiazole-2-amine,5-bromo-1,3,4-thiadiazole-2-ylamine,5-bromo-1,3,4 thiadiazol-2-ylamine,bromothiadiazolamine,pubchem20972,ksc496o1t PubChem CID: 3650883 IUPAC Name: 5-bromo-1,3,4-thiadiazol-2-amine SMILES: NC1=NN=C(Br)S1