missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-(4-pyridyl)-1,3,4-thiadiazole 98.0+%, TCI America™

Catalog No. A30381G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30381G 1 g
A30385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30381G Supplier TCI America Supplier No. A30381G

Chemical Identifiers

CAS 2002-04-2
Molecular Formula C7H6N4S
Molecular Weight (g/mol) 178.21
MDL Number MFCD00473692
InChI Key KTWDTPBHCWJWGJ-UHFFFAOYSA-N
Synonym 5-pyridin-4-yl-1,3,4-thiadiazol-2-amine, 2-amino-5-4-pyridyl-1,3,4-thiadiazole, 2-amino-5-4-pyridinyl-1,3,4-thiadiazole, 5-4-pyridyl-1,3,4-thiadiazol-2-amine, 5-pyridin-4-yl-1,3,4 thiadiazol-2-ylamine, 2-amino-5-pyridin-4-yl-1,3,4-thiadiazole, pyridine, 4-2-amino-1,3,4-thiadiazol-5-yl, 4-2-amino-1,3,4-thiadiazol-5-yl pyridine, 2-amino-5-4-piridil-1,3,4-tiadiazolo, 5-4-pyridyl-1,3,4-thiazol-2-yl-amine
PubChem CID 121269
IUPAC Name 5-pyridin-4-yl-1,3,4-thiadiazol-2-amine
SMILES C1=CN=CC=C1C2=NN=C(S2)N

Specifications

Melting Point 243°C
Color Yellow
Quantity 1 g
Formula Weight 178.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-(4-pyridyl)-1,3,4-thiadiazole
RTECSNumber US2168500
TSCA No