accessibility menu, dialog, popup

UserName

CAS RN 2002-04-2

IUPAC Name: 5-(pyridin-4-yl)-1,3,4-thiadiazol-2-amine
Synonyms: 5-(pyridin-4-yl)-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 178.21
Molecular Formula: C7H6N4S
InChi Key: KTWDTPBHCWJWGJ-UHFFFAOYSA-N
SMILES: NC1=NN=C(S1)C1=CC=NC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results