missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-(4-pyridyl)-1,3,4-thiadiazole, 97%

Catalog No. AAH2751403
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2751403 1 g
AAH2751409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2751403 Supplier Thermo Scientific Chemicals Supplier No. H2751403

CAS: 2002-04-2 | C9H6BrF3O | 267.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2002-04-2
MDL Number MFCD00473692

Specifications

Melting Point 242°C to 248°C
Molecular Formula C7H6N4S
Quantity 1 g
Synonym 5-pyridin-4-yl-1,3,4-thiadiazol-2-amine, 2-amino-5-4-pyridyl-1,3,4-thiadiazole, 2-amino-5-4-pyridinyl-1,3,4-thiadiazole, 5-4-pyridyl-1,3,4-thiadiazol-2-amine, 5-pyridin-4-yl-1,3,4 thiadiazol-2-ylamine, 2-amino-5-pyridin-4-yl-1,3,4-thiadiazole, pyridine, 4-2-amino-1,3,4-thiadiazol-5-yl, 4-2-amino-1,3,4-thiadiazol-5-yl pyridine, 2-amino-5-4-piridil-1,3,4-tiadiazolo, 5-4-pyridyl-1,3,4-thiazol-2-yl-amine
InChI Key KTWDTPBHCWJWGJ-UHFFFAOYSA-N
SMILES C1=CN=CC=C1C2=NN=C(S2)N
IUPAC Name 5-pyridin-4-yl-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol) 178.21
PubChem CID 121269
Formula Weight 178.21
Percent Purity 97%
Chemical Name or Material 2-Amino-5-(4-pyridyl)-1,3,4-thiadiazole
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber US2168500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only