missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(trifluoromethyl)pyrimidine 98.0+%, TCI America™

Catalog No. A25225G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25225G 5 g
A25221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25225G Supplier TCI America Supplier No. A25225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16075-42-6
Molecular Formula C5H4F3N3
Molecular Weight (g/mol) 163.10
MDL Number MFCD00828624
InChI Key NKOTXYPTXKUCDL-UHFFFAOYSA-N
Synonym 2-amino-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-amine, 2-amino-4-trifluoromethylpyrimidine, 2-pyrimidinamine, 4-trifluoromethyl, 4-trifluoromethyl-2-pyrimidinamine, 4-trifluoromethyl-pyrimidin-2-ylamine, 4-trifluoromethyl pyrimidin-2-ylamine, 4-trifluoromethyl pyrimidine-2-ylamine
PubChem CID 595915
IUPAC Name 4-(trifluoromethyl)pyrimidin-2-amine
SMILES NC1=NC=CC(=N1)C(F)(F)F

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 5 g
Formula Weight 163.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-(trifluoromethyl)pyrimidine