missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. p-7117774
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2168200MG 200 mg
A21681G 1 g
A21685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A2168200MG Supplier TCI America Supplier No. A2168200MG
Only null left

Chemical Identifiers

CAS 106447-97-6
Molecular Formula C6H5F3N2
Molecular Weight (g/mol) 162.115
MDL Number MFCD00077455
InChI Key RWGBXAQMUBGGKQ-UHFFFAOYSA-N
Synonym 2-amino-4-trifluoromethyl pyridine, 4-trifluoromethyl pyridin-2-amine, 4-trifluoromethyl-pyridin-2-ylamine, 2-amino-4-trifluoromethylpyridine, 4-trifluoromethyl-2-pyridinamine, 4-trifluoromethyl-2-pyridylamine, 2-pyridinamine, 4-trifluoromethyl, 2-amino-4-trifluoromethyl-pyridine, pubchem2977
PubChem CID 2778315
IUPAC Name 4-(trifluoromethyl)pyridin-2-amine
SMILES C1=CN=C(C=C1C(F)(F)F)N

Specifications

Melting Point 73°C
Color White-Yellow
Quantity 200 mg
Formula Weight 162.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-(trifluoromethyl)pyridine