accessibility menu, dialog, popup

UserName

CAS RN 106447-97-6

NFFFH2N

CAS RN 106447-97-6

IUPAC Name: 4-(trifluoromethyl)pyridin-2-amine
Synonyms: 4-(trifluoromethyl)pyridin-2-amine
Molecular Weight (g/mol): 162.12
Molecular Formula: C6H5F3N2
InChi Key: RWGBXAQMUBGGKQ-UHFFFAOYSA-N
SMILES: NC1=NC=CC(=C1)C(F)(F)F
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)