missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. p-7118005
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26481G 1 g
A26485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26481G Supplier TCI America Supplier No. A26481G

Chemical Identifiers

CAS 1483-54-1
Molecular Formula C8H5F3N2
Molecular Weight (g/mol) 186.137
MDL Number MFCD04974124
InChI Key IAIRNHIXDCZUCV-UHFFFAOYSA-N
Synonym 3-Amino-4-cyanobenzotrifluoride, 2-Cyano-5-(trifluoromethyl)aniline, 4-(Trifluoromethyl)anthranilonitrile
PubChem CID 13439028
IUPAC Name 2-amino-4-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C(F)(F)F)N)C#N

Specifications

Melting Point 90°C
Color Yellow
Boiling Point 127°C
Quantity 1 g
UN Number 3439
Formula Weight 186.14
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-(trifluoromethyl)benzonitrile