missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-thiazoleacetic acid, 97%

Catalog No. p-7027069
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2353414 25 g
AAB2353422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2353414 Supplier Thermo Scientific Chemicals Supplier No. B2353414
Only null left

CAS: 29676-71-9 | C5H6N2O2S | 158.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29676-71-9
Molecular Formula C5H6N2O2S
Molecular Weight (g/mol) 158.175
MDL Number MFCD00010068
InChI Key DYCLHZPOADTVKK-UHFFFAOYSA-N
Synonym 2-amino-4-thiazoleacetic acid, 2-2-aminothiazol-4-yl acetic acid, 2-aminothiazol-4-acetic acid, 2-aminothiazole-4-yl acetic acid, 4-thiazoleacetic acid, 2-amino, 2-amino-1,3-thiazol-4-yl acetic acid, 2-aminothiazole-4-yl-acetic acid, 2-aminothiazole-4-acetic acid, unii-qsh3o4tnjt, 2-2-amino-1,3-thiazol-4-yl acetic acid
PubChem CID 34665
IUPAC Name 2-(2-amino-1,3-thiazol-4-yl)acetic acid
SMILES C1=C(N=C(S1)N)CC(=O)O

Specifications

Melting Point ∼130°C (decomposition)
Quantity 25 g
Beilstein 127415
Formula Weight 158.18
Percent Purity 97%
Chemical Name or Material 2-Amino-4-thiazoleacetic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 249-769-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only