missing translation for 'onlineSavingsMsg'
Learn More

2-amino-4-picoline, 98%

Catalog No. AC153861000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153861000 100 g Glass bottle
AC153865000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153861000 Supplier Thermo Scientific Chemicals Supplier No. 153861000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 695-34-1 | C6H8N2 | 108.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 695-34-1
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006321
InChI Key ORLGLBZRQYOWNA-UHFFFAOYSA-N
Synonym 2-amino-4-methylpyridine, 2-amino-4-picoline, 2-pyridinamine, 4-methyl, ascensil, 4-methyl-2-pyridinamine, 4-methyl-2-pyridylamine, 4-methyl-2-aminopyridine, 4-picoline, 2-amino, w 45 raschig, aminton
PubChem CID 1533
IUPAC Name 4-methylpyridin-2-amine
SMILES CC1=CC(=NC=C1)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 96°C to 101°C
Color Beige to Yellow or Brown
Boiling Point 230°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 22, V,9, 325
Merck Index 15, 462
Solubility Information Solubility in water: 41g/L (20°C). Other solubilities: soluble in lower alcohols,slightly soluble in petroleum ether
Formula Weight 108.14
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material 2-Amino-4-picoline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Toxic in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-780-9

RUO – Research Use Only