missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-phenylthiazole 98.0+%, TCI America™

Catalog No. p-7118107
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28461G 1 g
A28465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28461G Supplier TCI America Supplier No. A28461G
Only null left

Chemical Identifiers

CAS 2010-06-2
Molecular Formula C9H8N2S
Molecular Weight (g/mol) 176.24
MDL Number MFCD00039680
InChI Key PYSJLPAOBIGQPK-UHFFFAOYSA-N
Synonym 4-phenylthiazol-2-amine, 2-amino-4-phenylthiazole, phenthiazamine, 2-thiazolamine, 4-phenyl, 4-phenyl-2-thiazolamine, thiazole, 2-amino-4-phenyl, 4-phenyl-thiazol-2-ylamine, 4-phenyl-2-thiazolylamine, 2-amino-4-phenyl-1,3-thiazole, 4-phenyl-2-aminothiazole
PubChem CID 40302
IUPAC Name 4-phenyl-1,3-thiazol-2-amine
SMILES C1=CC=C(C=C1)C2=CSC(=N2)N

Specifications

Melting Point 151°C
Quantity 1 g
UN Number 2811
Formula Weight 176.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-phenylthiazole
EINECSNumber (5)-0208
RTECSNumber XJ2878000
TSCA No