accessibility menu, dialog, popup

UserName

CAS RN 2010-06-2

IUPAC Name: 4-phenyl-1,3-thiazol-2-amine
Synonyms: phenthiazamine
Molecular Weight (g/mol): 176.24
Molecular Formula: C9H8N2S
InChi Key: PYSJLPAOBIGQPK-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)