missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-phenylthiazole, 98%

Catalog No. AAA1848803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1848803 1 g
AAA1848806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1848803 Supplier Thermo Scientific Chemicals Supplier No. A1848803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2010-06-2 | C9H8N2S | 176.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2010-06-2
Molecular Formula C9H8N2S
Molecular Weight (g/mol) 176.24
MDL Number MFCD00039680
InChI Key PYSJLPAOBIGQPK-UHFFFAOYSA-N
Synonym 4-phenylthiazol-2-amine, 2-amino-4-phenylthiazole, phenthiazamine, 2-thiazolamine, 4-phenyl, 4-phenyl-2-thiazolamine, thiazole, 2-amino-4-phenyl, 4-phenyl-thiazol-2-ylamine, 4-phenyl-2-thiazolylamine, 2-amino-4-phenyl-1,3-thiazole, 4-phenyl-2-aminothiazole
PubChem CID 40302
IUPAC Name 4-phenyl-1,3-thiazol-2-amine
SMILES C1=CC=C(C=C1)C2=CSC(=N2)N

Specifications

Melting Point 148°C to 152°C
Odor Odorless
Quantity 1 g
UN Number UN2811
Beilstein 128609
Formula Weight 176.24
Percent Purity 98%
Chemical Name or Material 2-Amino-4-phenylthiazole
missing translation for 'healthHazard1' H301-H315-H318-H335
missing translation for 'healthHazard2' GHS H Statement
H301
Toxic if swallowed.
missing translation for 'healthHazard3' P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 217-926-8
RTECSNumber XJ2878000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only