missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-phenylthiazole, 97%

Catalog No. AC432490050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432490050 5 g Glass bottle
AC432490010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432490050 Supplier Thermo Scientific Chemicals Supplier No. 432490050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2010-06-2 | C9H8N2S | 176.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2010-06-2
Molecular Formula C9H8N2S
Molecular Weight (g/mol) 176.24
MDL Number MFCD00039680
InChI Key PYSJLPAOBIGQPK-UHFFFAOYSA-N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 150.0°C to 152.5°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Synonym 2-benzyloxy-3-methoxybenzaldehyde, 2-benzyloxy-3-methoxy-benzaldehyde, 3-methoxy-2-phenylmethoxy benzaldehyde, acmc-1cljo, benzyloxy-3-methoxy benzaldehyde, 2-benzyloxy-3-methoxyl-benzaldehyde, 2-benzyloxy-3-methoxybenzenecarbaldehyde
SMILES NC1=NC(=CS1)C1=CC=CC=C1
IUPAC Name 3-methoxy-2-phenylmethoxybenzaldehyde
PubChem CID 1268087
Formula Weight 176.24
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 2-Amino-4-phenylthiazole
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 217-926-8

RUO – Research Use Only