missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-nitrophenol Sodium Salt 80.0+%, TCI America™

Catalog No. p-7116866
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A038025G 25 g
A0380500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A038025G Supplier TCI America Supplier No. A038025G
Only null left

Chemical Identifiers

CAS 61702-43-0
Molecular Formula C6H5N2NaO3
Molecular Weight (g/mol) 176.107
MDL Number MFCD00067428
InChI Key QROXVHUAKDVYQE-UHFFFAOYSA-M
Synonym 2-Hydroxy-5-nitroaniline Sodium Salt
PubChem CID 5485279
IUPAC Name sodium;2-amino-4-nitrophenolate
SMILES C1=CC(=C(C=C1[N+](=O)[O-])N)[O-].[Na+]

Specifications

Quantity 25 g
Formula Weight 176.11
Percent Purity ≥80.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-nitrophenol Sodium Salt
RTECSNumber SJ6305000
TSCA Yes