missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-nitrobenzoic acid, 97%

Catalog No. AC156980250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156980250 25 g Glass bottle
AC156980050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156980250 Supplier Thermo Scientific Chemicals Supplier No. 156980250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 619-17-0 | C7H6N2O4 | 182.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-17-0
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.14
MDL Number MFCD00007262
InChI Key UEALKTCRMBVTFN-UHFFFAOYSA-N
Synonym 2-amino-4-nitrobenzoic acid, 4-nitroanthranilic acid, benzoic acid, 2-amino-4-nitro, 2-amino-4-nitrobenzoicacid, anthranilic acid, 4-nitro, ccris 441, 2-amino-4-nitro-benzoic acid, 4-nitro-2-aminobenzoic acid, dsstox_cid_963, dsstox_rid_75891
PubChem CID 12076
ChEBI CHEBI:75335
IUPAC Name 2-amino-4-nitrobenzoic acid
SMILES NC1=CC(=CC=C1C(O)=O)[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 268°C to 269°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula NO2C6H3(NH2)CO2H
Quantity 25 g
Beilstein 14, 374
Formula Weight 182.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-nitrobenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-583-5

RUO – Research Use Only