missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-nitrobenzoic acid, 97+%

Catalog No. p-7056082
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6483114 25 g
AAH6483122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6483114 Supplier Thermo Scientific Chemicals Supplier No. H6483114

CAS: 619-17-0 | C7H6N2O4 | 182.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 619-17-0
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.14
MDL Number MFCD00007262
InChI Key UEALKTCRMBVTFN-UHFFFAOYSA-N
Synonym 2-amino-4-nitrobenzoic acid, 4-nitroanthranilic acid, benzoic acid, 2-amino-4-nitro, 2-amino-4-nitrobenzoicacid, anthranilic acid, 4-nitro, ccris 441, 2-amino-4-nitro-benzoic acid, 4-nitro-2-aminobenzoic acid, dsstox_cid_963, dsstox_rid_75891
PubChem CID 12076
ChEBI CHEBI:75335
IUPAC Name 2-amino-4-nitrobenzoic acid
SMILES NC1=CC(=CC=C1C(O)=O)[N+]([O-])=O

Specifications

Melting Point 257°C (literature)
Quantity 25 g
Beilstein 785071
Formula Weight 182.13
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 2-Amino-4-nitrobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-583-5
RTECSNumber CB3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only