missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylthiazole 99.0+%, TCI America™

Catalog No. p-7117361
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A130325G 25 g
A1303250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A130325G Supplier TCI America Supplier No. A130325G
Only null left

Chemical Identifiers

CAS 1603-91-4
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.166
MDL Number MFCD00005329
InChI Key OUQMXTJYCAJLGO-UHFFFAOYSA-N
Synonym 2-amino-4-methylthiazole, 4-methylthiazol-2-amine, 2-thiazolamine, 4-methyl, 2-amino-4-methylthiazol, 4-methyl-2-thiazolamine, 4-methyl-2-thiazolylamine, 2-amino-4-methyl-1,3-thiazole, normotiroide, thiazole, 2-amino-4-methyl, 4-methyl-2-aminothiazole
PubChem CID 74143
ChEBI CHEBI:39753
IUPAC Name 4-methyl-1,3-thiazol-2-amine
SMILES CC1=CSC(=N1)N

Specifications

Melting Point 45°C
Color Yellow
Boiling Point 232°C
Quantity 25 g
Formula Weight 114.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methylthiazole
EINECSNumber (5)-0203
TSCA No
Recommended Storage Refrigerator