missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-methylthiazole Hydrochloride 98.0+%, TCI America™
$52.19 - $76.43
Chemical Identifiers
| CAS | 6142-15-0 |
|---|---|
| Molecular Formula | C4H7ClN2S |
| Molecular Weight (g/mol) | 150.62 |
| MDL Number | MFCD00035214 |
| InChI Key | WUMMJVLSXMYDRW-UHFFFAOYSA-N |
| PubChem CID | 80240 |
| IUPAC Name | hydrogen 4-methyl-1,3-thiazol-2-amine chloride |
| SMILES | [H+].[Cl-].CC1=CSC(N)=N1 |
Chemical Identifiers
| 6142-15-0 | |
| 150.62 | |
| WUMMJVLSXMYDRW-UHFFFAOYSA-N | |
| hydrogen 4-methyl-1,3-thiazol-2-amine chloride |
| C4H7ClN2S | |
| MFCD00035214 | |
| 80240 | |
| [H+].[Cl-].CC1=CSC(N)=N1 |
Specifications
| 6142-15-0 | |
| White | |
| MFCD00035214 | |
| WUMMJVLSXMYDRW-UHFFFAOYSA-N | |
| hydrogen 4-methyl-1,3-thiazol-2-amine chloride | |
| 80240 | |
| ≥98.0% (HPLC,N) | |
| 2-Amino-4-methylthiazole Hydrochloride |
| 173°C | |
| C4H7ClN2S | |
| 5 g | |
| [H+].[Cl-].CC1=CSC(N)=N1 | |
| 150.62 | |
| 150.62 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0203
RTECSNumber : XJ2630000
TSCA : Yes