accessibility menu, dialog, popup

UserName

HClCH3SH2NN

CAS RN 6142-15-0

IUPAC Name: hydrogen 4-methyl-1,3-thiazol-2-amine chloride
Synonyms: hydrogen 2-thiazolamine, 4-methyl- chloride
Molecular Weight (g/mol): 150.62
Molecular Formula: C4H7ClN2S
InChi Key: WUMMJVLSXMYDRW-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CSC(N)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results