Learn More
2-Amino-4-methylpyridine, 98%
CAS: 695-34-1 | C6H8N2 | 108.144 g/mol
Supplier: Thermo Scientific Chemicals A1417736
Description
2-Amino-4-methylpyridine has been used in the synthesis of 2-amino-4-methylÂpyridinium 2-hyÂdroxyÂbenzoate. It is potent inhibitor of NOS2 (iNOS) in vitro. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-4-methylpyridine has been used in the synthesis of 2-amino-4-methylÂpyridinium 2-hyÂdroxyÂbenzoate. It is potent inhibitor of NOS2 (iNOS) in vitro. It is used as pharmaceutical intermediate.
Solubility
Soluble in water (41 mg/ml at 20°C), lower alcohols (slightly), chloroform, ethyl acetate, and DMF.
Notes
Hygroscopic. Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water. Store at -20°C.
Specifications
| 2-Amino-4-methylpyridine | |
| 97°C to 101°C | |
| 118°C (244°F) | |
| C6H8N2 | |
| 500 g | |
| 107066 | |
| 14,466 | |
| Soluble in water (41mg/ml at 20°C),lower alcohols (slightly),chloroform,ethyl acetate,and DMF. | |
| CC1=CC(=NC=C1)N | |
| 108.144 | |
| 108.14 |
| 695-34-1 | |
| 231°C | |
| Unpleasant | |
| MFCD00006321 | |
| UN2811 | |
| Hygroscopic | |
| 2-amino-4-methylpyridine, 2-amino-4-picoline, 2-pyridinamine, 4-methyl, ascensil, 4-methyl-2-pyridinamine, 4-methyl-2-pyridylamine, 4-methyl-2-aminopyridine, 4-picoline, 2-amino, w 45 raschig, aminton | |
| ORLGLBZRQYOWNA-UHFFFAOYSA-N | |
| 4-methylpyridin-2-amine | |
| 1533 | |
| 98% |
Chemical Identifiers
| 695-34-1 | |
| 108.144 | |
| ORLGLBZRQYOWNA-UHFFFAOYSA-N | |
| 1533 | |
| CC1=CC(=NC=C1)N |
| C6H8N2 | |
| MFCD00006321 | |
| 2-amino-4-methylpyridine, 2-amino-4-picoline, 2-pyridinamine, 4-methyl, ascensil, 4-methyl-2-pyridinamine, 4-methyl-2-pyridylamine, 4-methyl-2-aminopyridine, 4-picoline, 2-amino, w 45 raschig, aminton | |
| 4-methylpyridin-2-amine |
Safety and Handling
GHS H Statement
H301-H311-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
H301+H311-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 211-780-9
RTECSNumber : TJ5150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only