missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylphenol, 98%

CAS: 95-84-1 | C7H9NO | 123.16 g/mol

Supplier:  Thermo Scientific Chemicals A1954618

 View more versions of this product

Catalog No. AAA1954618


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-4-methylphenol
134°C to 138°C
MFCD00007699
606494
ZMXYNJXDULEQCK-UHFFFAOYSA-N
2-amino-4-methylphenol
7264
98%
95-84-1
C7H9NO
50 g
2-amino-p-cresol, 2-hydroxy-5-methylaniline, phenol, 2-amino-4-methyl, 3-amino-4-hydroxytoluene, 6-hydroxy-m-toluidine, 4-methyl-2-aminophenol, 5-methyl-2-hydroxyaniline, 2-amino-4-cresol, p-cresol, 2-amino, 2-amino-4-methyl-phenol
CC1=CC=C(O)C(N)=C1
123.16
123.16

Chemical Identifiers

95-84-1
123.16
ZMXYNJXDULEQCK-UHFFFAOYSA-N
7264
CC1=CC=C(O)C(N)=C1
C7H9NO
MFCD00007699
2-amino-p-cresol, 2-hydroxy-5-methylaniline, phenol, 2-amino-4-methyl, 3-amino-4-hydroxytoluene, 6-hydroxy-m-toluidine, 4-methyl-2-aminophenol, 5-methyl-2-hydroxyaniline, 2-amino-4-cresol, p-cresol, 2-amino, 2-amino-4-methyl-phenol
2-amino-4-methylphenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 202-457-3

RTECSNumber : SJ6078000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only