missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylbenzothiazole, 98%

Catalog No. AAA1082206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1082206 5 g
AAA1082214 25 g
AAA1082222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1082206 Supplier Thermo Scientific Chemicals Supplier No. A1082206

CAS: 1477-42-5 | C8H8N2S | 164.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1477-42-5
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00005793
InChI Key GRIATXVEXOFBGO-UHFFFAOYSA-N
Synonym 2-amino-4-methylbenzothiazole, 4-methylbenzo d thiazol-2-amine, 2-benzothiazolamine, 4-methyl, 4-methyl-2-aminobenzothiazole, 4-methyl-2-benzothiazolamine, benzothiazole, 2-amino-4-methyl, 4-methylbenzothiazol-2-ylamine, 2-amino-4-methyl benzothiazole, 4-methyl-2-aminobenzothiozole, unii-45v5f1b9z8
PubChem CID 15132
IUPAC Name 4-methyl-1,3-benzothiazol-2-amine
SMILES CC1=C2N=C(N)SC2=CC=C1

Specifications

Melting Point 137°C to 139°C
Quantity 5 g
Beilstein 129346
Sensitivity Moisture sensitive
Formula Weight 164.22
Percent Purity 98%
Chemical Name or Material 2-Amino-4-methylbenzothiazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 216-028-3
RTECSNumber DL2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only