accessibility menu, dialog, popup

UserName

CH3SH2NN

CAS RN 1477-42-5

IUPAC Name: 4-methyl-1,3-benzothiazol-2-amine
Synonyms: 2-benzothiazolamine, 4-methyl-
Molecular Weight (g/mol): 164.23
Molecular Formula: C8H8N2S
InChi Key: GRIATXVEXOFBGO-UHFFFAOYSA-N
SMILES: CC1=C2N=C(N)SC2=CC=C1
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)