missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylbenzonitrile 98.0+%, TCI America™

Catalog No. A24481G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24481G 1 g
A24485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24481G Supplier TCI America Supplier No. A24481G

Chemical Identifiers

CAS 26830-96-6
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00173706
InChI Key LGNVAEIITHYWCG-UHFFFAOYSA-N
Synonym 2-cyano-5-methylaniline, 3-amino-4-cyanotoluene, 2-amino-4-methyl-benzonitrile, 4-methyl-2-aminobenzonitrile, benzonitrile, 2-amino-4-methyl, 2-amino-4-methylbenzenecarbonitrile, 2-amino-4-methylbenzonitril, 6-cyano-m-toluidine, pubchem4661, 4-methylanthranilonitrile
PubChem CID 2801276
IUPAC Name 2-amino-4-methylbenzonitrile
SMILES CC1=CC=C(C#N)C(N)=C1

Specifications

Melting Point 95°C
Quantity 1 g
UN Number 3439
Formula Weight 132.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methylbenzonitrile