missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-methylbenzamide, Thermo Scientific™

$111.90 - $111.90

Chemical Identifiers

CAS 39549-79-6
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.18
MDL Number MFCD00221474
InChI Key RUHKZVAPXHIWJH-UHFFFAOYSA-N
Synonym 2-amino-4-methyl-benzamide, benzamide, 2-amino-4-methyl, pubchem4030, 2-azanyl-4-methyl-benzamide, benzamide,2-amino-4-methyl, benzamide, 2-amino-4-methyl-9ci
PubChem CID 2801474
IUPAC Name 2-amino-4-methylbenzamide
SMILES CC1=CC(N)=C(C=C1)C(N)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
TL00794DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
TL00794DA
1 g Amber Glass Bottle
Each for $111.90
 
Description

Description

Specifications

Chemical Identifiers

39549-79-6
150.18
RUHKZVAPXHIWJH-UHFFFAOYSA-N
2801474
CC1=CC(N)=C(C=C1)C(N)=O
C8H10N2O
MFCD00221474
2-amino-4-methyl-benzamide, benzamide, 2-amino-4-methyl, pubchem4030, 2-azanyl-4-methyl-benzamide, benzamide,2-amino-4-methyl, benzamide, 2-amino-4-methyl-9ci
2-amino-4-methylbenzamide

Specifications

39549-79-6
97%
MFCD00221474
1 g
RUHKZVAPXHIWJH-UHFFFAOYSA-N
2-amino-4-methylbenzamide
2801474
97%
97.0
C8H10N2O
Amber Glass Bottle
2-amino-4-methyl-benzamide, benzamide, 2-amino-4-methyl, pubchem4030, 2-azanyl-4-methyl-benzamide, benzamide,2-amino-4-methyl, benzamide, 2-amino-4-methyl-9ci
CC1=CC(N)=C(C=C1)C(N)=O
150.18
150.18
2-Amino-4-methylbenzamide
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS