missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methoxyphenol, 97%

Catalog No. p-7032314
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3372403 1 g
AAH3372406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3372403 Supplier Thermo Scientific Chemicals Supplier No. H3372403

CAS: 20734-76-3 | C7H9NO2 | 139.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20734-76-3
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD06616911
InChI Key TUADYTFWZPZZTP-UHFFFAOYSA-N
Synonym 2-amino-4-methoxy-phenol, 2-hydroxy-5-methoxyaniline, phenol, 2-amino-4-methoxy, 4-methoxy-2-aminophenol, 3-amino-4-hydroxyanisole, asinex-reag bas 13015559, zlchem 183, acmc-1cs0c, 2-azanyl-4-methoxy-phenol, phenol,2-amino-4-methoxy
PubChem CID 1419108
IUPAC Name 2-amino-4-methoxyphenol
SMILES COC1=CC(=C(C=C1)O)N

Specifications

Melting Point 135°C to 137°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 139.16
Percent Purity 97%
Chemical Name or Material 2-Amino-4-methoxyphenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 687-284-5
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only