missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methoxy-6-methyl-1,3,5-triazine 98.0+%, TCI America™

Catalog No. A15605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15605G 5 g
A156025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15605G Supplier TCI America Supplier No. A15605G

Chemical Identifiers

CAS 1668-54-8
Molecular Formula C5H8N4O
Molecular Weight (g/mol) 140.146
MDL Number MFCD00052764
InChI Key NXFQWRWXEYTOTK-UHFFFAOYSA-N
Synonym 2-amino-4-methoxy-6-methyl-1,3,5-triazine, 2-methyl-4-amino-6-methoxy-s-triazine, 1,3,5-triazin-2-amine, 4-methoxy-6-methyl, 2-amino-4-methyl-6-methoxy-1,3,5-triazine, unii-395vj06rc4, s-triazine, 2-amino-4-methoxy-6-methyl, 4-methoxy-6-methyl-1,3,5-triazine-2-ylamine, pubchem21415, acmc-1buty, dsstox_cid_21230
PubChem CID 15466
ChEBI CHEBI:83511
IUPAC Name 4-methoxy-6-methyl-1,3,5-triazin-2-amine
SMILES CC1=NC(=NC(=N1)OC)N

Specifications

Color Yellow
Quantity 5 g
Formula Weight 140.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-methoxy-6-methyl-1,3,5-triazine
RTECSNumber XY3193000
TSCA Yes