accessibility menu, dialog, popup

UserName

H3CONCH3NH2NN

CAS RN 1668-54-8

IUPAC Name: 4-methoxy-6-methyl-1,3,5-triazin-2-amine
Synonyms: 4-methoxy-6-methyl-1,3,5-triazin-2-amine
Molecular Weight (g/mol): 140.15
Molecular Formula: C5H8N4O
InChi Key: NXFQWRWXEYTOTK-UHFFFAOYSA-N
SMILES: COC1=NC(C)=NC(N)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results