missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-hydroxyquinoline hydrate, 97%, water <12%

Catalog No. p-7046006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1276003 1 g
AAL1276006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1276003 Supplier Thermo Scientific Chemicals Supplier No. L1276003
Only null left

CAS: 42712-64-1 | C9H8N2O | 160.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42712-64-1
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.18
MDL Number MFCD00052386
InChI Key LWGUCIXHBVVATR-UHFFFAOYSA-N
Synonym 2-aminoquinolin-4-ol, 4-quinolinol, 2-amino, 2-amino-4-hydroxyquinoline, 2-aminoquinolin-4 1h-one, 2-amino-4-1h-quinolinone, 2-amino-quinolin-4-ol, 2-amino-4 1h-quinolinone, 4 1h-quinolinone, 2-amino, 2-?aminoquinolin-?4-?ol, aminoquinolinol
PubChem CID 594793
IUPAC Name 2-amino-1H-quinolin-4-one
SMILES NC1=CC(=O)C2=CC=CC=C2N1

Specifications

Melting Point ∽298°C (decomposition)
Quantity 1 g
Beilstein 124785
Formula Weight 160.18 (Anhydrous)
Percent Purity 97%
Assay water ≈10%
Chemical Name or Material 2-Amino-4-hydroxyquinoline hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only