missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4'-fluorobenzophenone 98.0+%, TCI America™

Catalog No. p-7118058
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27601G 1 g
A27605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27601G Supplier TCI America Supplier No. A27601G

Chemical Identifiers

CAS 3800-06-4
Molecular Formula C13H10FNO
Molecular Weight (g/mol) 215.23
InChI Key FFFXIQFESQNINT-UHFFFAOYSA-N
PubChem CID 9837287
IUPAC Name (2-aminophenyl)-(4-fluorophenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)F)N

Specifications

Melting Point 129°C
Color Yellow
Quantity 1 g
Formula Weight 215.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4′-fluorobenzophenone