missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorothiophenol, 96%

CAS: 1004-00-8 | C6H6ClNS | 159.631 g/mol

$139.63 - $448.38

Chemical Identifiers

CAS 1004-00-8
Molecular Formula C6H6ClNS
Molecular Weight (g/mol) 159.631
MDL Number MFCD00792528
InChI Key NGIRMPARLVGMPX-UHFFFAOYSA-N
Synonym 2-amino-4-chlorothiophenol, 2-amino-4-chlorobenzene-1-thiol, 2-amino-4-chloro-benzenethiol, benzenethiol, 2-amino-4-chloro, 4-chloro-2-aminothiophenol, 5-chloro-2-mercaptoaniline, 2-amino-4-chloro thiophenol, 5-chloro-2-mercaptoaniline hydrochloride, benzenethiol, 2-amino-4-chloro-, hydrochloride, pubchem5427
PubChem CID 12000
IUPAC Name 2-amino-4-chlorobenzenethiol
SMILES C1=CC(=C(C=C1Cl)N)S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2564006
View Documents
Thermo Scientific Chemicals
B2564006
5 g
Each for $139.63
Only null left
 
AAB2564014
View Documents
Thermo Scientific Chemicals
B2564014
25 g
Each for $448.38
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1004-00-8
159.631
NGIRMPARLVGMPX-UHFFFAOYSA-N
12000
C1=CC(=C(C=C1Cl)N)S
C6H6ClNS
MFCD00792528
2-amino-4-chlorothiophenol, 2-amino-4-chlorobenzene-1-thiol, 2-amino-4-chloro-benzenethiol, benzenethiol, 2-amino-4-chloro, 4-chloro-2-aminothiophenol, 5-chloro-2-mercaptoaniline, 2-amino-4-chloro thiophenol, 5-chloro-2-mercaptoaniline hydrochloride, benzenethiol, 2-amino-4-chloro-, hydrochloride, pubchem5427
2-amino-4-chlorobenzenethiol

Specifications

1004-00-8
Unpleasant
MFCD00792528
UN1759
2-amino-4-chlorothiophenol, 2-amino-4-chlorobenzene-1-thiol, 2-amino-4-chloro-benzenethiol, benzenethiol, 2-amino-4-chloro, 4-chloro-2-aminothiophenol, 5-chloro-2-mercaptoaniline, 2-amino-4-chloro thiophenol, 5-chloro-2-mercaptoaniline hydrochloride, benzenethiol, 2-amino-4-chloro-, hydrochloride, pubchem5427
C1=CC(=C(C=C1Cl)N)S
159.631
159.64
2-Amino-4-chlorothiophenol
52°C to 59°C
C6H6ClNS
5 g
Air Sensitive
NGIRMPARLVGMPX-UHFFFAOYSA-N
2-amino-4-chlorobenzenethiol
12000
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only