missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorothiophenol, 96%

Catalog No. AAB2564006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2564006 5 g
AAB2564014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2564006 Supplier Thermo Scientific Chemicals Supplier No. B2564006

CAS: 1004-00-8 | C6H6ClNS | 159.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1004-00-8
Molecular Formula C6H6ClNS
Molecular Weight (g/mol) 159.631
MDL Number MFCD00792528
InChI Key NGIRMPARLVGMPX-UHFFFAOYSA-N
Synonym 2-amino-4-chlorothiophenol, 2-amino-4-chlorobenzene-1-thiol, 2-amino-4-chloro-benzenethiol, benzenethiol, 2-amino-4-chloro, 4-chloro-2-aminothiophenol, 5-chloro-2-mercaptoaniline, 2-amino-4-chloro thiophenol, 5-chloro-2-mercaptoaniline hydrochloride, benzenethiol, 2-amino-4-chloro-, hydrochloride, pubchem5427
PubChem CID 12000
IUPAC Name 2-amino-4-chlorobenzenethiol
SMILES C1=CC(=C(C=C1Cl)N)S

Specifications

Melting Point 52°C to 59°C
Odor Unpleasant
Quantity 5 g
UN Number UN1759
Sensitivity Air Sensitive
Formula Weight 159.64
Percent Purity 96%
Chemical Name or Material 2-Amino-4-chlorothiophenol
Hazard Category H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only