missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloropyridine 98.0+%, TCI America™

Catalog No. p-7117762
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A21525G 5 g
A215225G 25 g
2 options

Catalog No. A21525G

Supplier: TCI America A21525G

Only null left

Chemical Identifiers

CAS 19798-80-2
Molecular Formula C5H6ClN2
Molecular Weight (g/mol) 129.57
MDL Number MFCD04113820
InChI Key RQMWVVBHJMUJNZ-UHFFFAOYSA-O
Synonym 2-amino-4-chloropyridine, 4-chloro-pyridin-2-ylamine, 2-pyridinamine, 4-chloro, 4-chloro-2-pyridinamine, 4-chloro-2-pyridylamine, 2-amino-4-chloro pyridine, 4-chloro-2-pyridinylamine, 2-amino-4-chloro-pyridine, 4-chloro-2-aminopyridine, 4-chlorpyridin-2-amin
PubChem CID 581866
IUPAC Name 2-amino-4-chloropyridin-1-ium
SMILES NC1=CC(Cl)=CC=[NH+]1

Specifications

Melting Point 131°C
Color White-Yellow
Quantity 5 g
Formula Weight 128.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4-chloropyridine