accessibility menu, dialog, popup

UserName

NH2ClHN

CAS RN 19798-80-2

IUPAC Name: 2-amino-4-chloropyridin-1-ium
Synonyms: 2-amino-4-chloropyridin-1-ium
Molecular Weight (g/mol): 129.57
Molecular Formula: C5H6ClN2
InChi Key: RQMWVVBHJMUJNZ-UHFFFAOYSA-O
SMILES: NC1=CC(Cl)=CC=[NH+]1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)