missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chlorophenol 98.0+%, TCI America™

$50.33 - $374.68

Chemical Identifiers

CAS 95-85-2
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD00007694
InChI Key SWFNPENEBHAHEB-UHFFFAOYSA-N
Synonym 5-chloro-2-hydroxyaniline, phenol, 2-amino-4-chloro, fouramine py, 4-chloro-2-aminophenol, 2-hydroxy-5-chloroaniline, p-chloro-o-aminophenol, c.i. oxidation base 18, 2-amino-4-chloro-phenol, unii-2z43lea3dx, ccris 4579
PubChem CID 7265
IUPAC Name 2-amino-4-chlorophenol
SMILES NC1=CC(Cl)=CC=C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A028425G
View Documents
TCI America
A028425G
25 g
Each for $50.33
Only null left
 
A0284500G
View Documents
TCI America
A0284500G
500 g
Each for $374.68
Only null left
 
Description

Description

Specifications

Chemical Identifiers

95-85-2
143.57
SWFNPENEBHAHEB-UHFFFAOYSA-N
7265
NC1=CC(Cl)=CC=C1O
C6H6ClNO
MFCD00007694
5-chloro-2-hydroxyaniline, phenol, 2-amino-4-chloro, fouramine py, 4-chloro-2-aminophenol, 2-hydroxy-5-chloroaniline, p-chloro-o-aminophenol, c.i. oxidation base 18, 2-amino-4-chloro-phenol, unii-2z43lea3dx, ccris 4579
2-amino-4-chlorophenol

Specifications

95-85-2
C6H6ClNO
25 g
5-chloro-2-hydroxyaniline, phenol, 2-amino-4-chloro, fouramine py, 4-chloro-2-aminophenol, 2-hydroxy-5-chloroaniline, p-chloro-o-aminophenol, c.i. oxidation base 18, 2-amino-4-chloro-phenol, unii-2z43lea3dx, ccris 4579
NC1=CC(Cl)=CC=C1O
143.57
143.57
Crystalline Powder
139°C
MFCD00007694
2673
SWFNPENEBHAHEB-UHFFFAOYSA-N
2-amino-4-chlorophenol
7265
≥98.0% (GC,T)
2-Amino-4-chlorophenol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0906

RTECSNumber : SJ5700000

TSCA : Yes

SDS
Documents

Documents