missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-chlorophenol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 139°C |
| Quantity | 500 g |
| UN Number | 2673 |
| Formula Weight | 143.57 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Amino-4-chlorophenol |
Safety and Handling
| EINECSNumber | (3)-0906 |
| RTECSNumber | SJ5700000 |
| TSCA | Yes |