missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4'-chlorodiphenyl Ether 98.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 202°C |
| Quantity | 5 g |
| Formula Weight | 219.67 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Amino-4′-chlorodiphenyl Ether |
Safety and Handling
| TSCA | No |