missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4'-chlorobiphenyl 98.0+%, TCI America™

Catalog No. A30345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30345G 5 g
A30341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30345G Supplier TCI America Supplier No. A30345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1204-44-0
Molecular Formula C12H10ClN
Molecular Weight (g/mol) 203.669
MDL Number MFCD03424659
InChI Key JPBWZIPCMDZOPM-UHFFFAOYSA-N
Synonym 4′-Chloro-[1,1′-biphenyl]-2-amine
PubChem CID 262261
IUPAC Name 2-(4-chlorophenyl)aniline
SMILES C1=CC=C(C(=C1)C2=CC=C(C=C2)Cl)N

Specifications

Melting Point 47°C
Quantity 5 g
Formula Weight 203.67
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4′-chlorobiphenyl