missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-chlorobenzonitrile 98.0+%, TCI America™
$108.11 - $333.67
Chemical Identifiers
| CAS | 38487-86-4 |
|---|---|
| Molecular Formula | C7H5ClN2 |
| Molecular Weight (g/mol) | 152.581 |
| MDL Number | MFCD00035927 |
| InChI Key | UZHALXIAWJOLLR-UHFFFAOYSA-N |
| Synonym | benzonitrile, 2-amino-4-chloro, 5-chloro-2-cyanoaniline, 4-chloro-2-aminobenzonitrile, 4-chloroanthranilonitrile, 2-amino-4-chlorobenzenecarbonitrile, pubchem20756, 2-amino-4-chloroberizonitrile, bidd:gt0102, 2-amino-4-chlorobenzonitrile, 2-amino-4-chloro-benzonitrile |
| PubChem CID | 170059 |
| IUPAC Name | 2-amino-4-chlorobenzonitrile |
| SMILES | C1=CC(=C(C=C1Cl)N)C#N |
Chemical Identifiers
| 38487-86-4 | |
| 152.581 | |
| UZHALXIAWJOLLR-UHFFFAOYSA-N | |
| 170059 | |
| C1=CC(=C(C=C1Cl)N)C#N |
| C7H5ClN2 | |
| MFCD00035927 | |
| benzonitrile, 2-amino-4-chloro, 5-chloro-2-cyanoaniline, 4-chloro-2-aminobenzonitrile, 4-chloroanthranilonitrile, 2-amino-4-chlorobenzenecarbonitrile, pubchem20756, 2-amino-4-chloroberizonitrile, bidd:gt0102, 2-amino-4-chlorobenzonitrile, 2-amino-4-chloro-benzonitrile | |
| 2-amino-4-chlorobenzonitrile |
Specifications
| 163°C | |
| Yellow | |
| Crystal-Powder at 20°C | |
| MFCD00035927 | |
| benzonitrile, 2-amino-4-chloro, 5-chloro-2-cyanoaniline, 4-chloro-2-aminobenzonitrile, 4-chloroanthranilonitrile, 2-amino-4-chlorobenzenecarbonitrile, pubchem20756, 2-amino-4-chloroberizonitrile, bidd:gt0102, 2-amino-4-chlorobenzonitrile, 2-amino-4-chloro-benzonitrile | |
| C1=CC(=C(C=C1Cl)N)C#N | |
| 152.581 | |
| 152.58 | |
| 2-Amino-4-chlorobenzonitrile |
| 38487-86-4 | |
| 5 g | |
| C7H5ClN2 | |
| 3439 | |
| UZHALXIAWJOLLR-UHFFFAOYSA-N | |
| 2-amino-4-chlorobenzonitrile | |
| 170059 | |
| ≥98.0% (GC) |
Safety and Handling
TSCA : Yes